Cole-Parmer® SHM-200 Series Stirring/Heating Mantles
Provides powerful stirring speeds up to 520 rpm and heating up to 450°C
  • Bidirectional stirring with stir bar auto capture function
  • Thermal insulating cartridge provides maximum heat transfer
  • Chemical resistant polypropylene housing is "safe to the touch"
Stir your sample while heating up to 450°C (842°F). These durable stirring/heating mantles feature a bidirectional magnetic stirrer with speeds up to 520 rpm. An “auto-recapture” function reactivates stirring if it is interrupted or lost for any reason.
A built-in power controller with dial markings from 1 to 10 offer great repeatability. Two colored lights indicate "power on" and "heat on." The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
Unique ventilation slots and ceramic fiber insulation keep the chemical-resistant polypropylene exterior cool even while in use. A safety earth (ground) screen and double fuses provide added safety.
£645.12 - £972.18GBP / Each
£774.14 - £1,166.62GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Power (VAC)
Availability Pricing
COMPARE
50 230
£645.12GBP / Each
£774.14GBP (incl VAT)
COMPARE
100 230
£660.03GBP / Each
£792.04GBP (incl VAT)
COMPARE
250 230
£701.19GBP / Each
£841.43GBP (incl VAT)
COMPARE
500 230
£732.27GBP / Each
£878.72GBP (incl VAT)
COMPARE
1000 230
£768.34GBP / Each
£922.01GBP (incl VAT)
COMPARE
2000 115
£972.18GBP / Each
£1,166.62GBP (incl VAT)
COMPARE
2000 230
£972.18GBP / Each
£1,166.62GBP (incl VAT)
Showing 1- 7 of 7