This product is now discontinued, View Related Products
Edwards Direct-Drive Rotary Vane Vacuum Pump
Rapid pumping speed with quiet operation and ability to handle vapor
  • Self-cooling motor shuts off if overheated
  • Air-tight chamber prevents oil suckback
  • Check oil condition levels through the sight glass
  • EN61010, CSA C22.2 1010, and UL61010 compliant
The E2M1.5 series dual-stage rotary vane vacuum pumps are rugged, mechanical, oil-sealed pumps that feature speeds of 1.6 m3/hr. Compact and quiet, they feature advanced lubrication circuits and high reliability—ideal for lab and light industrial applications.
Easy to install, the pumps come equipment with an integral IEC connector. The single-phase motor is enclosed and cooled by the motor-cooling fan. A thermal overload device protects the motor from overheating and automatically resets and restarts the pump once it cools down. The air-tight chamber features an oil distribution valve to prevent oil discharge into the pump interior after it stops. Easily inspect the oil levels through the sight glass.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Free Air Capacity (CFM) Power (VAC)
Availability Pricing
Edwards Direct-drive rotary vane vacuum pump, dual stage, 1.3 cfm, 115 VAC WZ-79300-20 Mfr # A37122902P (E2M1.5)
COMPARE
1.3 115
Call For Price
Edwards Direct-drive rotary vane vacuum pump, dual stage, 1.3 cfm, 230 VAC WZ-79300-25 Mfr # A37122919P (E2M1.5)
COMPARE
1.3 230