This product is now discontinued, View Related Products
Economical Type K/J Thermocouple Thermometers
Compact and lightweight design
  • Available with single or dual input
These thermometers provide accurate and reliable temperature measurement in an economic and compact design. Features include min/max, Hold, °F/°C selection, relative temperature, and auto power-off after 30 minutes of inactivity to save on battery power. Accept both type K or J thermocouple probes with a miniconnector. Available in single and dual input, with results displayed on a large 1 1/4" x 1 3/4" LCD. The dual input meter (94461-12) can read and display results from two probes simultaneously and track the difference between them (T1-T2). Order optional probes and other accessories separately.
What's Included: beaded wire probe (two probes for 94461-12) and one 9 V battery.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Data Logging Number Of Probe Channels Types Of Probes Accepted
Availability Pricing
General Tools DT80-1 Economical Type K/J Thermocouple Thermometer, Single Input WZ-94461-11 Mfr # DT80-1
COMPARE
1 Type J, Type K
Call For Price
General Tools DT80-2 Economical Type K/J Thermocouple Thermometer, Dual Input WZ-94461-12 Mfr # DT80-2
COMPARE
2 Type J, Type K