This product is now discontinued, View Related Products

Ecom Instruments Accessories for Wall Thickness Gauge

Probes suitable for thickness ranges from 0.06" to 15.7"
These probes are compatible with the 59785-19 wall thickness gauge. Gauge sold separately.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Probe Range
Availability Pricing
Ecom Instruments A0001503 Low Frequency Thickness Gauge Probe WZ-59785-01 Mfr # A0001503
COMPARE
Low-frequency 0.03\" to 15.7\" (0.7 to 400 mm)
Call For Price
Ecom Instruments A0001506 Miniature Thickness Gauge Probe WZ-59785-03 Mfr # A0001506
COMPARE
Mini 0.03\" to 15.7\" (0.7 to 400 mm)
Call For Price
Ecom Instruments A0001511 Standard Thickness Gauge Probe WZ-59785-05 Mfr # A0001511
COMPARE
Standard 0.03\" to 15.7\" (0.7 to 400 mm)
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat