BriskHeat® Dual-Element Heating Tapes with Benchtop Temperature Controller - Celsius - 240VAC
Convenient heating system that includes on/off temperature controller, flexible heating tape, and temperature sensor
  • Celsius controller temperature range 0 to 600°C
  • Factory-installed 5-foot J-Type thermocouple sensor
The flexible dual-element heating tape wraps around and heats a wide array of tubing, vessels, and apparatuses. The extremely flexible and high-temperature heating tape is made of two heating elements that are knitted together in series and then wound together with high-temperature Samox fiberglass.
The included digital on/off temperature controller provides basic control and connects to the heating tape without any additional wiring. The large display is easy to read and simple to program. Accuracy is +/- 1% FS. Hysteresis is adjustable between 1 and 99 degrees. In the case of probe failure, heating can be selected on, off, or periodical running. It is benchtop-friendly with its compact 5.5” x 4.25” x 1.75” profile. The controller has a 10amp fused output and can be used with other heating tapes, beaker heaters, and mantle heaters as long as they are equipped with standard U.S. plugs.
Approvals and Compliances: Controller: CE
Heating Tape: CE, RoHS, REACH
Warning: Heating tapes should not be overlapped.
£381.50 - £460.70GBP / Each
£457.80 - £552.84GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° C) Power (VAC)
Availability Pricing
BriskHeat CPSDCJCA-BWH052020LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 24" , 240 V WZ-36003-49 Mfr # CPSDCJCA-BWH052020LD
COMPARE
24 600 240
£381.50GBP / Each
£457.80GBP (incl VAT)
BriskHeat CPSDCJCA-BWH052040LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 48" , 240 V WZ-36003-51 Mfr # CPSDCJCA-BWH052040LD
COMPARE
48 600 240
£394.90GBP / Each
£473.88GBP (incl VAT)
BriskHeat CPSDCJCA-BWH052060LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 72" , 240 V WZ-36003-53 Mfr # CPSDCJCA-BWH052060LD
COMPARE
72 600 240
£414.40GBP / Each
£497.28GBP (incl VAT)
BriskHeat CPSDCJCA-BWH052080LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 96" , 240 V WZ-36003-55 Mfr # CPSDCJCA-BWH052080LD
COMPARE
96 600 240
£431.50GBP / Each
£517.80GBP (incl VAT)
BriskHeat CPSDCJCA-BWH052100LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1/2" x 120" , 240 V WZ-36003-57 Mfr # CPSDCJCA-BWH052100LD
COMPARE
120 600 240
£460.70GBP / Each
£552.84GBP (incl VAT)
BriskHeat CPSDCJCA-BWH102020LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1" x 24" , 240 V WZ-36003-59 Mfr # CPSDCJCA-BWH102020LD
COMPARE
24 600 240
£402.20GBP / Each
£482.64GBP (incl VAT)
BriskHeat CPSDCJCA-BWH102040LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1" x 48" , 240 V WZ-36003-61 Mfr # CPSDCJCA-BWH102040LD
COMPARE
48 600 240
£409.88GBP / Each
£491.86GBP (incl VAT)
BriskHeat CPSDCJCA-BWH102060LD Dual-Element Heating Tape with Temperature Controller, 0 to 600C, 1" x 6" , 240 Volt WZ-36003-63 Mfr # CPSDCJCA-BWH102060LD
COMPARE
6 600 240
£416.90GBP / Each
£500.28GBP (incl VAT)
Showing 1- 8 of 8