BriskHeat® Dual-Element Heating Tapes - 120 VAC
Greater flexibility than other high-temperature heating tapes
  • Heat to 1400°>F (760°>C) ideal for high-temperature use on conductive surfaces
Dual-element design improves the heater’s flexibility over standard high-temperature
tapes. Tapes are made of two heating elements that are knitted together in series and then
wound together with high-temperature Samox fiberglass. The 120 VAC tapes include a
plug
Warning: Heating tapes should not be overlapped and require the use of a temperature or
power controller sold separately.
£110.50 - £194.51GBP / Each
£132.60 - £233.41GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Width (in)
Availability Pricing
BriskHeat 051100LD Dual Element Heating Tape, 1/2" x 10ft, 780 Watts, 120V with 2-Prong Plug WZ-03106-26 Mfr # TBIH051-100LD
COMPARE
10 1400 1/2
£151.95GBP / Each
£182.34GBP (incl VAT)
BriskHeat 051120LD Dual Element Heating Tape, 1/2" x 12ft, 936 Watts, 120V with 2-Prong Plug WZ-03106-27 Mfr # TBIH051-120LD
COMPARE
12 1400 1/2
£184.09GBP / Each
£220.91GBP (incl VAT)
BriskHeat 101060LD Dual Element Heating Tape, 1" x 6ft, 936 Watts, 120V with 2-Prong Plug WZ-03106-28 Mfr # TBIH101-060LD
COMPARE
6 1400 1
£194.51GBP / Each
£233.41GBP (incl VAT)
BriskHeat TBIH051-020LD Dual Element, Cloth Insulated Heat Tape, 2' x 1/2" , 156 Watts, 120 V WZ-36050-00 Mfr # TBIH051-020LD
COMPARE
2 1400 1/2
£110.50GBP / Each
£132.60GBP (incl VAT)
BriskHeat TBIH052-020LD Dual Element, Cloth Insulated Heat Tape, 2' x 1/2" , 156 Watts, 240 V WZ-36050-05 Mfr # TBIH052-020LD
COMPARE
2 1400 1/2
£129.39GBP / Each
£155.27GBP (incl VAT)
BriskHeat TBIH051-040LD Dual Element, Cloth Insulated Heat Tape, 4' x 1/2" , 312 Watts, 120 V WZ-36050-10 Mfr # TBIH051-040LD
COMPARE
4 1400 1/2
£145.99GBP / Each
£175.19GBP (incl VAT)
BriskHeat TBIH051-060LD Dual Element, Cloth Insulated Heat Tape, 6' x 1/2" , 468 Watts, 120 V WZ-36050-20 Mfr # TBIH051-060LD
COMPARE
6 1400 1/2
£155.70GBP / Each
£186.84GBP (incl VAT)
BriskHeat TBIH051-080LD Dual Element, Cloth Insulated Heat Tape, 8' x 1/2" , 624 Watts, 120 V WZ-36050-30 Mfr # TBIH051-080LD
COMPARE
8 1400 1/2
£144.29GBP / Each
£173.15GBP (incl VAT)
BriskHeat TBIH101-020LD Dual Element, Cloth Insulated Heat Tape, 2' x 1" , 312 Watts, 120 V WZ-36050-60 Mfr # TBIH101-020LD
COMPARE
2 1400 1
£124.29GBP / Each
£149.15GBP (incl VAT)
BriskHeat TBIH101-040LD Dual Element, Cloth Insulated Heat Tape, 4' x 1" , 624 Watts, 120 V WZ-36050-70 Mfr # TBIH101-040LD
COMPARE
4 1400 1
£126.10GBP / Each
£151.32GBP (incl VAT)
Showing 1- 10 of 10