DWK Wheaton® Dounce Tissue Grinders, Glass
Reduce soft tissue to primary cells and organelles
  • Made from low extractable borosilicate glass
  • Two capacities available to suit your sample size
These grinders are designed to retain a high percentage of cell nuclei and mitochondria in soft tissues, such as egg, embryo, animal organ, or plant. In operation, the loose fitting pestle ball is encircled in liquid, which avoids heat build-up. Mortar has large reservoir and pouring lip. Each mortar is supplied with a “loose” pestle and a “tight” pestle.
£359.21 - £395.45GBP / Case of 2
£431.05 - £474.54GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Overall Length (mm)
Availability Pricing
DWK Life Sciences (Wheaton) 357538 Dounce Tissue Grinders, Glass, 1 mL; 2/Cs WZ-04468-00 Mfr # 357538
COMPARE
1 Glass 125
£359.21GBP / Case of 2
£431.05GBP (incl VAT)
DWK Life Sciences (Wheaton) 357542 Dounce Tissue Grinders, Glass, 7 mL; 2/Cs WZ-04468-02 Mfr # 357542
COMPARE
7 Glass 175
£395.45GBP / Case of 2
£474.54GBP (incl VAT)