This product is now discontinued, View Related Products
DispoTube Disposable Liquid Samplers
Ergonomic handle provides a good grip when sampling
  • Two way sampling—all layer and target point
  • Cleanroom manufactured
  • Individually packaged
Simply insert an open-top DispoTube sampler into the liquid to take an all-layer sample; or insert while closed then open to obtain a target-point sample. These single-use HDPE samplers are manufactured in a cleanroom and individually packaged to prevent contamination ensuring more accurate results.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
Dynalon DispoTube Samplers, 50 cm L, 100 mL, HDPE; Pk of 20 WZ-33687-66 Mfr # 853933-6620
COMPARE
19.625 DispoTube Samplers, 50 cm L, 100 mL, HDPE; Pk of 20
Call For Price
Dynalon DispoTube Samplers, Sterile, 50 cm L, 100 mL, HDPE; Pk of 20 WZ-33687-67 Mfr # 853933-6621
COMPARE
19.625 DispoTube Samplers, Sterile, 50 cm L, 100 mL, HDPE; Pk of 20
Call For Price
Dynalon DispoTube Samplers, 100 cm L, 200 mL, HDPE; Pk of 20 WZ-33687-68 Mfr # 853933-6630
COMPARE
39.375 DispoTube Samplers, 100 cm L, 200 mL, HDPE; Pk of 20
Call For Price
Dynalon DispoTube Samplers, Sterile, 100 cm L, 200 mL, HDPE; Pk of 20 WZ-33687-69 Mfr # 853933-6631
COMPARE
39.375 DispoTube Samplers, Sterile, 100 cm L, 200 mL, HDPE; Pk of 20