Cole-Parmer® Essentials Disposable Transfer Pipettes
Easily draw and deliver precise solutions
  • Deliver repeatable, uniform drops
  • Eliminate cross contamination with these low cost, disposable LDPE pipettes
Use these unbreakable low-density polyethylene (LDPE) pipettes to eliminate the chance of contamination from rubber bulbs. They deliver repeatable, uniform drops and are configured to fit into a variety of vessels. Choose the bulb draw and graduations or drops/mL needed for measurement and delivery.
£66.23 - £97.52GBP / Box of 500
£79.48 - £117.02GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Overall Length (in) Description
Availability Pricing
COMPARE
1.5 4.625 Transfer Pipette, Graduated, Small Bulb, 1.5 mL; 500/Pk
£66.23GBP / Box of 500
£79.48GBP (incl VAT)
COMPARE
15 6.125 Transfer Pipette, Jumbo-bulb, Long-Stem, 15.0 mL; 250/Pk
£68.20GBP / Box of 250
£81.84GBP (incl VAT)
COMPARE
7.5 6.125 Transfer Pipette, Graduated, Large Bulb, 7.5 mL; 500/Pk
£68.20GBP / Box of 500
£81.84GBP (incl VAT)
COMPARE
5.8 6.25 Transfer Pipette, Fine-tip, 5.8 mL; 500/Pk
£97.52GBP / Box of 500
£117.02GBP (incl VAT)
COMPARE
6 9 Transfer Pipette, Extra-Long Stem, 6 mL; 400/Pk
£88.01GBP / Box of 500
£105.61GBP (incl VAT)