This product is now discontinued, View Related Products
Bürkle Disposable Open-Tip Piercing Lance Samplers
Easily take samples directly from the vessel with the durable piercing tip
  • Closed- and open-tip options available for different sampling methods
  • Manufactured in a cleanroom ISO Class 7 environment and individually packaged
  • Certified FDA 21 CFR 5 compliant for food applications
These HDPE samplers are ideal for taking large and small samples of powder and granulate directly from the bag. Choose from either sterile or nonsterile configurations. The sterilized options are sterilized via gamma irradiation.
The open-tip sampler is suitable for free-flowing samples, which flow through the open tube and directly into a container or bag that is held underneath. The volume of sample taken is variable, and dependent on the sampling container used at the end of the sampler.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Sterile Description
Availability Pricing
Burkle 5393-5560 Disposable Piercing Sampling Lance, HDPE; 300 mm x 14 mm WZ-06043-42 Mfr # 5393-5560
COMPARE
11.8 No Disposable piercing sampling lance, HDPE, 300 mm x 14 mm
Call For Price
Burkle 5393-5561 Disposable Piercing Sampling Lance, HDPE, 300 mm x 14 mm; Sterile WZ-06043-43 Mfr # 5393-5561
COMPARE
11.8 Yes Disposable piercing sampling lance, HDPE, 300 mm x 14 mm, sterile
Call For Price
Burkle 5393-5570 Disposable Piercing Sampling Lance, HDPE; 500 mm x 14 mm WZ-06043-44 Mfr # 5393-5570
COMPARE
19.7 No Disposable piercing sampling lance, HDPE, 500 mm x 14 mm
Call For Price
Burkle 5393-5571 Disposable Piercing Sampling Lance, HDPE, 500 mm x 14 mm; Sterile WZ-06043-45 Mfr # 5393-5571
COMPARE
19.7 Yes Disposable piercing sampling lance, HDPE, 500 mm x 14 mm, sterile