This product is now discontinued, View Related Products
Disposable Cross-Section Powder Sampling Lances
A perfect sample every time
  • Visually confirm sample quality with the open-channel design
  • Manufactured in a cleanroom and individually packaged
  • Choose between gamma sterilized and nonsterile lances
This disposable sampling lance withdraws an all-layer cross-section and is ideally suited to powders and other slightly cohesive solids. Sturdy HDPE construction and beveled tip easily pierce compacted powders. All lances are manufactured in a cleanroom environment and individually wrapped. Select the gamma-sterilized version for an additional level of purity.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Sterile Volume (mL)
Availability Pricing
Burkle Disposable Cross-Section Powder Sampling Lance, 300 mm x 21 mm, 60 mL; 20/pk WZ-06043-31 Mfr # 5393-5510
COMPARE
11.811 No 60
Call For Price
Burkle Sterile Disposable Cross-Section Powder Sampling Lance, 300 mm x 21 mm, 60 mL; 20/pk WZ-06043-32 Mfr # 5393-5511
COMPARE
11.811 Yes 60
Call For Price
Burkle Disposable Cross-Section Powder Sampling Lance, 500 mm x 21 mm, 100 mL; 20/pk WZ-06043-33 Mfr # 5393-5521
COMPARE
19.685 No 100
Call For Price
Burkle Sterile Disposable Cross-Section Powder Sampling Lance, 500 mm x 21 mm, 100 mL; 20/pk WZ-06043-34 Mfr # 5393-5522
COMPARE
19.685 Yes 100