This product is now discontinued, View Related Products
DispoPicker Disposable Solids Samplers
Don't let your sample flow back into the container
  • Closed tip prevents sample from trickling out of the shaft during withdrawal
  • Open-channel design allows you to see your sample
  • Powders and small-grain bulk materials work best
This HDPE solids sampler features a closed tip to hold the sample within the shaft while withdrawing and transferring. The stability of the sample ensures a representative cross-section is taken.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
Dynalon DispoPicker Open Channel Solids Sampler w/ Closed Tip, 60 mL, 300 mm L; 20/pk WZ-33687-39 Mfr # 853933-5540
COMPARE
11.875 Open Channel Solids Sampler w/ Closed Tip, 60 mL, 300 mm L; 20/pk
Call For Price
Dynalon DispoPicker Sterile Open Channel Solids Sampler w/ Closed Tip, 60 mL, 300 mm L; 20/pk WZ-33687-41 Mfr # 853933-5541
COMPARE
11.875 Sterile Open Channel Solids Sampler w/ Closed Tip, 60 mL, 300 mm L; 20/pk
Call For Price
Dynalon DispoPicker Open Channel Solids Sampler w/ Closed Tip, 100 mL, 500 mm L; 20/pk WZ-33687-43 Mfr # 853933-5550
COMPARE
19.625 Open Channel Solids Sampler w/ Closed Tip, 100 mL, 500 mm L; 20/pk
Call For Price
Dynalon DispoPicker Sterile Open Channel Solids Sampler w/ Closed Tip, 100 mL, 500 mm L; 20/pk WZ-33687-45 Mfr # 853933-5551
COMPARE
19.625 Sterile Open Channel Solids Sampler w/ Closed Tip, 100 mL, 500 mm L; 20/pk