This product is now discontinued, View Related Products
Digital Solar-Powered Industrial Pressure Gauge
The first-ever "solar only" digital pressure gauge
  • Large display
  • <10 Lux rating- only one foot-candle
  • User selectable functions
  • IP65 weatherproof housing
  • ASME B40.7 conformance
Threre's no need to ever change a battery or push an on button with these solar-powered digital industrial gauges. The special features of this gauge make it an incredible low-cost solution for many industrial applications. The gauge can run on very low light–<10 Lux rating–that's only one foot-candle!
Call For Price
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Min Pressure (PSI) Process Connection
Availability Pricing
Weiss DUGY1-0015-4L Pressure Gauge Vacuum-15 PSI, 1/4" NPT(M); pHosphor Bronze Sensor/brass Socket WZ-68900-25 Mfr # DUGY1-0015-4L
COMPARE
15 -15 1/4\" NPT(M)
Call For Price
Weiss DUGY1-0060-4L Pressure Gauge Vacuum-60 PSI, 1/4" NPT(M); pHosphor Bronze Sensor/brass Socket WZ-68900-27 Mfr # DUGY1-0060-4L
COMPARE
60 -15 1/4\" NPT(M)
Call For Price
Weiss DUGY1-0100-4L Pressure Gauge Vacuum-100 PSI, 1/4" NPT(M); pHosphor Bronze Sensor/brass Socket WZ-68900-29 Mfr # DUGY1-0100-4L
COMPARE
100 -15 1/4\" NPT(M)
Call For Price
Weiss DUGY1-0200-4L Pressure Gauge 0-200 PSI, 1/4" NPT(M); pHosphor Bronze Sensor/brass Socket WZ-68900-31 Mfr # DUGY1-0200-4L
COMPARE
200 0 1/4\" NPT(M)
Call For Price
Weiss DUGY1-0400-4L Pressure Gauge 0-400 PSI, 1/4" NPT(M); pHosphor Bronze Sensor/brass Socket WZ-68900-33 Mfr # DUGY1-0400-4L
COMPARE
400 0 1/4\" NPT(M)
Call For Price
Weiss DUGY1-1000-4L Pressure Gauge 0-1000 PSI, 1/4" NPT(M); pHosphor Bronze Sensor/brass Socket WZ-68900-35 Mfr # DUGY1-1000-4L
COMPARE
1000 0 1/4\" NPT(M)
Call For Price
Weiss DUGY2-0015-4L Pressure Gauge Vacuum-15 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-37 Mfr # DUGY3-015-4L
COMPARE
15 -15 1/4\" NPT(M)
Call For Price
Weiss DUGY2-0060-4L Pressure Gauge Vacuum-60 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-39 Mfr # DUGY3-060-4L
COMPARE
60 -15 1/4\" NPT(M)
Call For Price
Weiss DUGY2-0100-4L Pressure Gauge Vacuum-100 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-41 Mfr # DUGY3-100-4L
COMPARE
100 -15 1/4\" NPT(M)
Call For Price
Weiss DUGY2-0200-4L Pressure Gauge 0-200 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-43 Mfr # DUGY3-200-4L
COMPARE
200 0 1/4\" NPT(M)
Call For Price
Weiss DUGY2-0400-4L Pressure Gauge 0-400 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-45 Mfr # DUGY3-400-4L
COMPARE
400 0 1/4\" NPT(M)
Call For Price
Weiss DUGY2-1000-4L Pressure Gauge 0-1000 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-47 Mfr # DUGY3-1000-4L
COMPARE
1000 0 1/4\" NPT(M)
Call For Price
Weiss DUGY2-0015-2L Pressure Gauge Vacuum-15 PSI, 1/2" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-49 Mfr # DUGY3-015-2L
COMPARE
15 -15 1/2\" NPT(M)
Call For Price
Weiss DUGY2-0060-2L Pressure Gauge Vacuum-60 PSI, 1/2" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-51 Mfr # DUGY3-060-2L
COMPARE
60 -15 1/2\" NPT(M)
Call For Price
Weiss DUGY2-0100-2L Pressure Gauge Vacuum-100 PSI, 1/2" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-53 Mfr # DUGY3-100-2L
COMPARE
100 -15 1/2\" NPT(M)
Call For Price
Weiss DUGY2-0200-2L Pressure Gauge 0-200 PSI, 1/2" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-55 Mfr # DUGY3-200-2L
COMPARE
200 0 1/2\" NPT(M)
Call For Price
Weiss DUGY2-0400-2L Pressure Gauge 0-400 PSI, 1/2" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-57 Mfr # DUGY3-400-2L
COMPARE
400 0 1/2\" NPT(M)
Call For Price
Weiss DUGY2-1000-2L Pressure Gauge 0-1000 PSI, 1/2" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-59 Mfr # DUGY3-1000-2L
COMPARE
1000 0 1/2\" NPT(M)
Call For Price
Weiss Pressure Gauge 0-3000 PSI, 1/2" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-61 Mfr # DUGY2-3000-2L
COMPARE
3000 0 1/2\" NPT(M)
Call For Price
Weiss Pressure Gauge 0-5000 PSI, 1/2" NPT(M); 316 SS Sensor/ 316 SS Socket WZ-68900-63 Mfr # DUGY2-5000-2L
COMPARE
5000 0 1/2\" NPT(M)
Call For Price
Showing 1 - 20 of 21