General Tools Digital Manometer with Backlight Display
Read gauge and differential pressure in 11 units of measure!
  • Ideal for measuring low differential pressure in clean rooms, test and balance, HVAC, pneumatic systems, and computer peripherals
  • Large backlight LCD, maximum, minimum, hold, and auto power off functions
  • Built-in RS-232 port downloads information to your PC
  • Order cable and software separately from Accessories below
These manometers are for use with noncorrosive gases and air only, not liquids.
£312.73GBP / Each
£375.28GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (PSID) Resolution
Availability Pricing
General Tools DM8215 Digital Manometer with range of 0 to 15 psi WZ-68603-02 Mfr # DM8215
COMPARE
±0.3% full-scale 15 0.01 psi
£312.73GBP / Each
£375.28GBP (incl VAT)
General Tools DM8200 Digital Manometer with range of 0 to 100 psi WZ-68603-06 Mfr # DM8200
COMPARE
±0.3% full-scale 100 0.1 psi
£312.73GBP / Each
£375.28GBP (incl VAT)