Digi-Sense Thermowells - Brass
Designed for easy replacement of your industrial probe without shutting the operation down.
To match your probe to a thermowell, first measure the distance from the probe’s tip to the top of the thread (typical thread length is 0.5" for 1/2" NPT fittings). Next, match this length to the probe depth of the thermowell (“A” dimension). Other thermowell sizes and materials are available—call our Application Specialists for more information.
Limited-Space Thermowell when you need a shorter insertion depth.
Standard Thermowell for general-purpose applications.
Heavy-Duty Thermowell for higher fluid velocity ratings.
£19.65 - £47.32GBP / Each
£23.58 - £56.78GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in) Material
Availability Pricing
COMPARE
Brass
£19.65GBP / Each
£23.58GBP (incl VAT)
COMPARE
Brass
£43.53GBP / Each
£52.24GBP (incl VAT)
COMPARE
Brass
£47.32GBP / Each
£56.78GBP (incl VAT)