Digi-Sense Thermocouple Hook-And-Loop Strap-On Surface Probes
Designed for Easy Installation & Removal on Round or Oddly Shaped Vessels
  • Hook-and-loop strap makes for easy installation and removal
  • Strap can be used for permanent or temporary installation
  • Response Time: 1.25 seconds or greater
  • Each probe comes with color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
£59.99 - £63.95GBP / Each
£71.99 - £76.74GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
212 J
£63.95GBP / Each
£76.74GBP (incl VAT)
COMPARE
212 T
£59.99GBP / Each
£71.99GBP (incl VAT)