This product is now discontinued, View Related Products
Digi-Sense Temperature Probe Compression Fittings 6544
Fittings let you adjust the depth your probe extends into a system for temperature readings. Use fittings for straight-shaft probes and metal tubing only. Tighten compression nut to secure and completely seal probe.
Replacement ferrules let you reuse a compression fitting when replacing a worn-out probe. Simply disassemble fitting, discard probe and old ferrule, install replacement ferrule over probe sheath, and retighten fitting.
Call For Price
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Length (in) Wire Gauge (Ga)
Availability Pricing
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
Showing 1- 11 of 11