Digi-Sense® Special-Purpose RTD Probes with Braided Cables
Designed for long term applications or unique environment requirements
  • Time Constant: 3 seconds or greater
  • Custom designs or changes to this product are available upon request
Maximum error for probe assembly is ±1.1°C + 0.12% of reading below 300°F, ±0.35% of reading above 300°F. Alpha coefficient is 0.003850 omega/omega/°C (DIN IEC 751). Resistance at ice point (0°C) is 100 omega.
Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five.
Note the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C) are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
£95.45 - £153.19GBP / Each
£114.54 - £183.83GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Description
Availability Pricing
COMPARE
900 Alligator Clip RTD Probe, 100 Ohm, Digi-Sense® 3-Pin Connector, 1.39\" Length, 10 ft Stainless Steel Braid Cable
£153.19GBP / Each
£183.83GBP (incl VAT)
COMPARE
900 Alligator Clip RTD Probe, 100 Ohm, ANSI 3-Blade Connector, 1.39\" Length, 10 ft Stainless Steel Braid Cable
£146.51GBP / Each
£175.81GBP (incl VAT)
COMPARE
900 Compact RTD Probe, 100 Ohm, ANSI 3-Blade Connector, 1.5\" Length, 5 ft Stainless Steel Braid Cable
£95.45GBP / Each
£114.54GBP (incl VAT)