Digi-Sense RTD Solar Twin Temp Industrial Probes
An innovative, cost effective way to solve two types of temperature measuring needs while still only using one entry point into your process.
Cole Parmer solar Twin Temp industrial thermometers are the perfect solution for systems requiring local and remote temperature readings. Local readings are provided by digital screen. The remote reading is achieved with a continuou 4-20 mA output signal. All sensors are located in the thermometer stem and will easily fit into existing bi-metal thermowells of corresponding length. The housing is manufactured of 304 stainless and is guaranteed to be hermetically sealed to prevent fogging.
£537.33GBP / Each
£644.80GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
COMPARE
1/2\" NPT(M) 300
£537.33GBP / Each
£644.80GBP (incl VAT)