This product is now discontinued, View Related Products
Digi-Sense Advanced Benchtop Temperature Controllers
Outlet receptacle allows for direct plug-in of heating devices
  • Advanced design allows for quicker and more reliable results
  • Improved display with easy-to-use interface
The Digi-Sense temperature controllers can be used for a wide variety of applications. Provides excellent control accuracy and power capabilities, making them ideal for pilot process plants, R & D labs, or for OEM requirements.
Product FeaturesAccepts eight thermocouple types with miniconnectors.
General features—Rear panel output receptacle for direct plug-in of heaters and other resistive devices. Field calibrations help to improve system accuracy by entering the offset value to correct for individual probe error. Simultaneously view both the measured and set point on the two-line alphanumeric display. Temperature scale selectable to read in °F, °C, K (Kelvin), or °R (Rankine). Front panel indicates output and alarm conditions. Multiple control modes from simple On/Off control to sophisticated autotuning PID control.
Safety features—Output power to load device is automatically shut down in the event of a broken sensor or control loop break. Overtemp feature protection shuts down the system if the user- settable over-temperature or timer is exceeded. Outlet power receptacle is fused separately from the controller for added safety. Audible and visual out-of-range alarms are also included.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
115 TC-9000 Thermocouple Temperature Controller, 115V
Call For Price
COMPARE
230 TC-9000 Thermocouple Temperature Controller, 230V
Call For Price
COMPARE
115 TC-9500 Multiparameter Temperature Controller with USB Output, 115V
Call For Price
COMPARE
230 TC-9500 Multiparameter Temperature Controller with USB Output, 230V