This product is now discontinued, View Related Products
Dickson Temperature and Humidity Dataloggers 61625
Record up to 32,512 temperature and humidity data points
  • Push-to-start models give you control in data recording
  • Provides 21 CFR Part 11 compliance with optional software
  • Use software to calculate dew point and to create custom graphs
  • of your data
Use these dataloggers to monitor food, chemical, drug, and equipment storage areas, clean rooms, incubators, and more! Units offer a push-to-start function that gives the operator more control of data recording. The easy to read LCD display allows you to view current and min/max readings with the push of a button and the red LED flashes to confirm sampling. The compact rugged design allows you to place these dataloggers just about anywhere.
The Windows® based software lets you set sampling intervals, view downloaded data, and create customized graphs and tables in seconds. It can even plot data from several loggers on a single graph to let you compare conditions. Software is required to program dataloggers; order separately below.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
Dickson SP425 USB Temperature Datalogger WZ-18010-00 Mfr # SP425
COMPARE
158 ±1.2 1
Call For Price
Dickson TP425 USB Temperature/Humidity Datalogger WZ-18010-01 Mfr # TP425
COMPARE
158 ±0.8 (over range +20 to +120); ±1.8 (< +20 and > +120) 1