Dibafit™ PEEK Multiport Mountable Block Connectors
Use with Omnifit® gripper fittings or Omni-Lok™ Type P/S for an inert fluid path with zero dead volume
  • Constructed of durable PEEK
These block connectors have 1/4"-28 UNF(F) threads and 1.5 mm internal bore. Select from 3-way or 4-way configuration.
£47.64GBP / Each
£57.17GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Thread size UNF(F) (port 1) (in) Description
Availability Pricing
Dibafit™ Multiport Connector, 3-Way, 1/4"-28 UNF(F) flat bottom, PEEK™, mountable; 1/ea WZ-21939-11 Mfr # 001057
COMPARE
1/4\"-28 Dibafit™ Multiport Connector, 3-Way, 1/4\"-28 UNF(F) flat bottom, PEEK™, mountable; 1/ea
£47.64GBP / Each
£57.17GBP (incl VAT)