Dibafit™ In-Line Coupling with 1/4"-28 UNF(F) Connector
Compact in size—ideal when space is limited
  • Constructed of durable PEEK
Use these PEEK in-line couplings to quickly and easily connect tubing lines. First tubing line passes through the connector and is held securely in place by a gripper ferrule. The 1/4"-28 UNF(F) port accepts a standard threaded fitting from a second line.
£99.10GBP / Pkg of 10
£118.92GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Orifice Dimensions (in)
Availability Pricing
Dibafit™ In-Line Connector, 1/4"-28 UNF(F) flat bottom, 1/8" OD tubing; 10/pk WZ-21940-06 Mfr # 002334
COMPARE
1/8
£99.10GBP / Pkg of 10
£118.92GBP (incl VAT)