This product is now discontinued, View Related Products
Dibafit™ 1/4"-28 UNF(M) Fittings for Flared Tubing
Create a ferrule- and leak-free connection while minimizing dead-volume
  • Constructed of durable PEEK
Dibafit™ Fittings for Flared Tubing provide a seal with only the tube material in the fluid path. A seal is created by compressing the flared portion of the tube against the flat bottom of the port. Use flanging kit 21940-23 to easily flange semi-rigid tubing. A washer is recommended to ensure the flare seals correctly. An O-ring and a second washer may be necessary to ensure the fitting seats correctly in deeper ports.
£214.88GBP / Pkg of 10
£257.86GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Compression Size / Tube Od (Port 2) (in) Fitting Material
Availability Pricing
Dibafit™ Flanged Tubing End Fitting, 1/4"-28 UNF(M), 1/16" OD flared tubing; 10/pk WZ-21939-73 Mfr # 002123
COMPARE
Natural 1/16 PEEK
Call For Price
Dibafit™ Flanged Tubing End Fitting, 1/4"-28 UNF(M), 1/8" OD flared tubing; 10/pk WZ-21939-91 Mfr # 002223
COMPARE
Natural 1/8 PEEK
£214.88GBP / Pkg of 10
£257.86GBP (incl VAT)