This product is now discontinued, View Related Products
Desco Surface Resistance Checker
Easily determine if a surface is dissipative, conductive, or insulative
  • Ensure compliance to ANSI/ESD S20.20-2007 per ESD TR53
  • Use optional electrodes for resistance measurements of larger surfaces
This portable meter is perfect for quick checks of surface resistance for ESD control applications in electronics manufacturing or handling environments. Simply press and hold the test button until the bright LEDs display the reading level. Unit features an automatic test voltage selector allowing the test voltage to switch from 10 V to 100 V if the measured resistance exceeds 1 x 10E5 ohms. Two banana jacks and an electrode toggle switch allow for the connection of two optional external 5-lb electrodes that measure surface resistance point to point, or resistance to equipment ground.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Accuracy Power (VAC) Range
Availability Pricing
Desco 19640 Surface Resistance Checker with NIST Calibration Certificate WZ-20028-01 Mfr # 19640
COMPARE
103 to 1012Ω
Call For Price
Desco 19641 Test Leads for Surface Resistance Checker, 1 Pair WZ-20028-03 Mfr # 19641
COMPARE
Call For Price
Desco 50003 Five Pound Electrodes for Surface Resistance Checker WZ-20028-05 Mfr # 50003