This product is now discontinued, View Related Products
Desco Static Control Wrist Straps
Protect delicate electronics from static damage
Wristbands and straps have 4-mm stainless steel snap studs. Purchase with or without a coiled cord; cords are 6-ft long and include a 1-MΩ resistor. Cleanroom compatible and safe for microelectronics applications. One size fits all. UL-listed.
Speidel® Twist-O-Flex® Wristbands feature a 300-series stainless steel interior expansion band. Flexible design won't pinch or grab hair.
Elastic Adjustable Wristbands feature a wide, soft elastic band and 300-series stainless steel used for all metal parts.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Accuracy Power (VAC) Range
Availability Pricing
Desco 09041 Adjustable Metal Speidel Wristband without Cord WZ-26809-00 Mfr # 09041
COMPARE
Call For Price
Desco 09085 Adjustable Metal Speidel Wristband with 6' Cord WZ-26809-02 Mfr # 09085
COMPARE
Call For Price
Desco 09028 Adjustable Elastic Wristband without Cord WZ-26809-04 Mfr # 09028
COMPARE
Call For Price
Desco 09070 Adjustable Elastic Wristband with 6' Cord WZ-26809-06 Mfr # 09070
COMPARE
Call For Price
Desco 09029 Replacement Adjustable Elastic Strip for 09028/09070 WZ-26809-08 Mfr # 09029
COMPARE
Call For Price
Desco 09480 Replacement 6' Black Coil Cord WZ-26809-09 Mfr # 09480
COMPARE
Call For Price
Desco 09680 Replacement 12' Black Coil Cord WZ-26809-10 Mfr # 09680
COMPARE
Call For Price
Showing 1- 7 of 7