This product is now discontinued, View Related Products
DESCO Form-Fitting ESD Gloves
Ideal for tactile sensitivity when handling small parts
These form-fitting nylon gloves eliminate static charges, ensuring superior protection when handling ESD-sensitive devices. Durable elastic band provides good ventilation and minimizes sweat buildup. Gloves provide an RTG of 105 to 108 Ω and are tested per ANSI/ESD 5P15.1 Standard Practice for In-Use Resistance Testing of Gloves and Finger Cots. Gloves come in pairs.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Size
Availability Pricing
Desco 68120 Form-Fitting ESD Gloves, Small, 7 3/8" (18.7 cm) WZ-26808-80 Mfr # 68120
COMPARE
7 3/8 Small
Call For Price
Desco 68121 Form-Fitting ESD Gloves, Medium, 7 1/2" (19.1 cm) WZ-26808-81 Mfr # 68121
COMPARE
7 1/2 Medium
Call For Price
Desco 68122 Form-Fitting ESD Gloves, Large, 7 3/4" (19.7 cm) WZ-26808-82 Mfr # 68122
COMPARE
7 3/4 Large
Call For Price
Desco 68123 Form-Fitting ESD Gloves, X-Large, 8" (20.3 cm) WZ-26808-83 Mfr # 68123
COMPARE
8 X-Large