DeFelsko PosiTest DFT Coating Thickness Gauges - All Metal Substrates
Quickly measure coating thickness on all metal substrates
  • Fast, repeatable measurements
  • No calibration required for most applications
  • Flip display for right-side-up viewing in any position
  • Zero feature for rough or curved surfaces
  • Handy reset feature when no zero reference is available
  • Strong, wear-resistant, ruby-tipped probe
  • Audible and visible measurement indication
  • V-groove in probe for positioning on cylindrical parts
  • Built-in wrist strap for added convenience and security
  • Mils/Microns switchable
  • Basic instructions on the back of each gauge
  • Small, simple, lightweight
  • Conforms to international standards including ASTM, ISO and others
These PosiTest DFT coating thickness gauges are perfect for providing quick, repeatable measurements on metal substrates. Calibration is not required for most applications making the the gauges ideal for powder coaters, paint applicators, coating inspectors, automotive resellers/refinishers, and more.
Units provide audible and visual measurement indications with a zero feature for rough or curved surfaces. The strong, wear-resistant, ruby-tipped probe is encased in an ergonomically designed body that includes a flip display allowing right-side-up viewing in any position.
Additional features include switchable unit of measure (mils/microns), reset function when no zero reference is available, and built-in wrist strap for added convenience and security.
Gauges conform to: ISO 2178/2360/2808, prEN ISO 19840, ASTM 244/B499/B659/D1186/D1400/D7091-05 E376/G12, BS3900-C5, SSPC-PA2 and more
£450.75GBP / Each
£540.90GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Range Resolution Scale
Availability Pricing
DeFelsko PosiTest DFT Ferrous Digital Fe Coating Gauge Thickness Gauge WZ-59785-13 Mfr # DFTF-C
COMPARE
0 to 40 mil (0 to 1000 µm) 0.1 mil (2.0 µm) Ferrous
£450.75GBP / Each
£540.90GBP (incl VAT)