Data Loggers With Data View Software
Model L322
  • 2 independent input channels
  • -20 to +20mAdc
  • Programmable storage rates from 8 per second to 1 per day
  • 3 user selectable storage modes
  • Stores up to 240,000 measurements in non-volatile memory
  • Scaling and engineering units inputted through software prior to saving
  • Powered by standard Alkaline batteries
  • Lightweight, compact, fits anywhere
  • 5 LED indicators quickly and clearly display logger status
  • 50V Cat. I
    Model L562
  • 2 input channels
  • Voltage: 0 to 600Vac/dc TRMS
  • Current: compatible with current probes with voltage outputs (see page 13)
  • 64 samples per cycle
  • 3 user selectable storage modes
  • Programmable storage rates from 8 per second to 1 per day
  • Stores up to 240,000 measurements in non-volatile memory
  • Powered by standard Alkaline batteries
  • Lightweight, compact, fits anywhere
  • 5 LED indicators quickly and clearly display logger status
  • 300V Cat. IV; 600V Cat. III with a safety rated current probe attached
  • £440.23GBP / Each
    £528.28GBP (incl VAT)
    1 variations of this product are available.
    See All Product Options Below
    1 PRODUCT OPTIONS
    Compareitem specifications
    Input Type Max Current (AAC) Max Voltage (VAC)
    Availability Pricing
    AEMC L652 TRMS Voltage/Current Model L562 WZ-26060-11 Mfr # 2126.35
    COMPARE
    Voltage and current 1 600
    £440.23GBP / Each
    £528.28GBP (incl VAT)