This product is now discontinued, View Related Products

Danger: Keep Out Signs

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): Indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MATR111VS Safety Sign, Danger Keep Out, 7 X 10, Adhesive Vinyl WZ-40710-06 Mfr # MATR111VS
COMPARE
Vinyl
Call For Price
Accuform MATR111VP Safety Sign, Danger Keep Out, 7 X 10, Plastic WZ-40710-07 Mfr # MATR111VP
COMPARE
Plastic
Call For Price
Accuform MATR111VA Safety Sign, Danger Keep Out, 7 X 10, Aluminum WZ-40710-08 Mfr # MATR111VA
COMPARE
Aluminum
Call For Price
Accuform MADM064VS Safety Sign, Danger - Keep Out, 10" X 14", Adhesive Vinyl WZ-41014-06 Mfr # MADM064VS
COMPARE
10 Adhesive Vinyl 14
Call For Price
Accuform MADM064VP Safety Sign, Danger - Keep Out, 10" X 14", Plastic WZ-41014-07 Mfr # MADM064VP
COMPARE
10 Plastic 14
Call For Price
Accuform MADM064VA Safety Sign, Danger - Keep Out, 10" X 14", Aluminum WZ-41014-08 Mfr # MADM064VA
COMPARE
10 Aluminum 14
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat