Culture Bottle Roller Stacking Platforms
Save lab counter space by stacking rollers
  • Allows placement of additional roller inside incubator
  • Constructed of cast acrylic and aluminum
This raised platform is designed to fit inside an incubator to allow for stacking of one or two Low Profile rollers inside.
Constructed of cast acrylic and aluminum for years of service inside high humidity, CO2 incubators without discoloring or rusting.
£323.47GBP / Each
£388.16GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Description
Availability Pricing
COMPARE
9 3/8 Cast Acrylic and Aluminum Culture Bottle Roller Stacking Platform; Single
£323.47GBP / Each
£388.16GBP (incl VAT)