This product is now discontinued, View Related Products
Crowcon Gasman Single-Gas Monitor
Datalog and download up to 3000 hours with time and date stamp
  • Rechargeable lithium-ion battery for 12 hours of operation
  • Rugged housing is IP 65/67 rated for dust and water resistance
Gasman monitor is available in a variety of toxic gas configurations to meet your individual application. Small size and light weight ensure the monitors are comfortable to wear all day long. Visible flashing LED, audible 85 dB, and vibrating alarms quickly alert the wearer to potential gas hazards. Data is recorded by the monitor and can be downloaded to a PC or printer using the adapter/PC interface (sold separately).
What's included: calibration cap, pocket clip, and rechargeable lithium-ion battery. (Order adapter/PC interface separately.)
Approval: UL and cUL Class I Div. 1 Groups A, B, C, D (pending), ATEX, CE
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Gases Measured
Availability Pricing
Crowcon GASMAN-OX Single-gas Monitor, O2 WZ-86550-00 Mfr # GASMAN-OX
COMPARE
O2
Call For Price
Crowcon GASMAN-CO Single-Gas Monitor, CO WZ-86550-02 Mfr # GASMAN-CO
COMPARE
CO
Call For Price
Crowcon GASMAN-H2S Single-gas Monitor, H2S WZ-86550-04 Mfr # GASMAN-H2S
COMPARE
H2S
Call For Price
Crowcon GASMAN-FL Single-gas Monitor, Lel (combustibles) WZ-86550-06 Mfr # GASMAN-FL
COMPARE
Combustibles
Call For Price
Crowcon GASMAN-SO2 Single-gas Monitor, So2 WZ-86550-08 Mfr # GASMAN-SO2
COMPARE
SO2
Call For Price
Crowcon GASMAN-NO Single-Gas Monitor, NO WZ-86550-10 Mfr # GASMAN-NO
COMPARE
Nitric oxide
Call For Price
Crowcon GASMAN-NO2 Single-gas Monitor, No2 WZ-86550-12 Mfr # GASMAN-NO2
COMPARE
Nitrogen dioxide
Call For Price
Crowcon GASMAN-O3 Single-gas Monitor, Ozone WZ-86550-14 Mfr # GASMAN-O3
COMPARE
Ozone
Call For Price
Showing 1- 8 of 8