Corning® Starter XT Ice-Free Coolers
No mess, no maintenance
  • Keep samples cold without melting ice contamination, inconvenient batteries, or power cord clutter
  • Cooling core maintains sample temperatures at 0.5°C to 4°C for at least four hours
  • Compatible with all Corning CoolSink® and CoolRack® sample modules (order separately)
These benchtop coolers eliminate possible contamination from ice, maintenance costs associated with battery replacement, and messy cords. Simply freeze the cooling core in a -20°C freezer for 12 or more hours, then remove and wait for core to reach 1°C before placing in holder. Once the core is installed, add your CoolSink or CoolRack module and load your samples. Samples quickly equilibrate to the core temperature and remain chilled for 4 to 16 hours.
Choose the Starter system with a low profile and smaller footprint (ideal for use in hoods) for shorter-term cooling, up to four hours.
£254.26GBP / Each
£305.11GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color
Availability Pricing
Corning XT Starter Ice-Free Cooler Holder with Cooling Core WZ-04392-14 Mfr # 432014
COMPARE
Purple / Gray
£254.26GBP / Each
£305.11GBP (incl VAT)