Compact Threaded Diaphragm Check Valves, 1/4" NPT(F)
Self-centering sealing disc ensures reliable sealing every time
  • Leak-free sealing prevents reverse flow
  • Create a secure seal using pipe thread connections
  • Requires only 1/2 psi of back pressure to close
  • Opens with minimal pressure and maintains a tight, consistent seal
  • Constant flushing action of the inlet flow keeps the sealing area clean
  • 1/4" NPT(F) x 1/4" NPT(F) connections
The compact diaphragm check valves use a flexible elastomer disc to deliver reliable, low-pressure sealing. In addition, the disc responds to extremely minimal pressure, allowing the valve to open easily. The self-centering design of the disc ensures that it seals in the exact same position every time, improving reliability and extending service life. As inlet flow passes through the valve, it continuously flushes the disc, keeping the sealing surface clean and free of particles, minimizing the risk of leakage.
Note: Check valves depend on gravity for sealing in low flow applications. Install check valves so gravity allows the disc to sit against the valve seat when flow is not present.
£187.10 - £648.50GBP / Each
£224.52 - £778.20GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Housing Material Inlet Port Outlet Port
Availability Pricing
Compact Check Valve, PP, 1/4" NPT(F) 4852711 Mfr # CKD025V-PP
COMPARE
Polypropylene 1/4" NPT(F) 1/4" NPT(F)
£215.90GBP / Each
£259.08GBP (incl VAT)
Compact Check Valve, PVC, 1/4" NPT(F) 4852712 Mfr # CKD025V-PV
COMPARE
PVC 1/4" NPT(F) 1/4" NPT(F)
£187.10GBP / Each
£224.52GBP (incl VAT)
Compact Check Valve, PVDF, 1/4" NPT(F) 4852713 Mfr # CKD025V-PF
COMPARE
PVDF 1/4" NPT(F) 1/4" NPT(F)
£648.50GBP / Each
£778.20GBP (incl VAT)