This product is now discontinued, View Related Products
Combination Wrench Sets
Compact reversing lever switches direction without removing from fastener
  • Reversing feature requires only a 5° handle swing compared to 30° on standard wrenches
  • Ratcheting box end is angled 15° to provide better fastening strength
  • Open-end jaws surfaces are parallel for better griping and fastening
  • Surface drive design nearly eliminates fastener rounding
  • Meets all ANSI and ASME specifications
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Wrench Type
Availability Pricing
COMPARE
SAE
Call For Price
COMPARE
Metric