This product is now discontinued, View Related Products
Comark® Food Service Waterproof Folding Thermometer with Bluetooth®
Wirelessly transfer temperature measurements via Bluetooth
  • Ideal for use in foodservice and HACCP programs
  • Eliminate the need for handwritten measurements with app and Bluetooth connection
  • Advanced features include Max, Min, and Hold functions
  • Exceptional battery life of up to 1000 hours
  • Pocket-size thermometer with folding probe—convenient for carrying
  • Adjust probe angle to go where you need it
This food thermometer is easy to use — simply swing probe open at any angle for ergonomic use. Now with Bluetooth® connectivity, you can wirelessly transmit the temperature readings right to your smartphone or tablet. Use with the Comark Kitchen Check App to quickly manage, review, report, and analyze your readings.
Fold up adjustable 4⅓” (110mm) stainless steel probe for best measurement and viewing angle.
The protective housing with rubber boot is IP65 rated to handle demanding environments. Plus, the BioCote® antimicrobial surface protection eliminated concerns of cross-contamination. BioCote® is an anti-microbial agent that is impregnated into the case during molding. Effectively inhibits the growth of micro-organisms and is an accepted part of HACCP standards.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Display Type Probe Length (in) Description
Availability Pricing
COMARK BT125KC Folding Food Service Thermometer with Bluetooth; °C WZ-90025-51 Mfr # BT125KC
COMPARE
°C LCD 4 21/64 Folding Food Service Thermometer with Bluetooth; °C
Call For Price
COMARK BT250KC Folding Food Service Thermometer with Bluetooth; °F WZ-90025-52 Mfr # BT250KC
COMPARE
°F LCD 4 21/64 Folding Food Service Thermometer with Bluetooth; °F