This product is now discontinued, View Related Products
Cole-Parmer Vertical Slab Class Plate Sets
Glass plate set consists of a notched plate and a back plate of 1/8'' soda lime glass. Radius-bottom corners on the back plate accept Gel Wrap™ gaskets for a leakproof seal. All edges are sanded and polished.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Glass Height (cm) Glass Width (cm) Description
Availability Pricing
COMPARE
14.5 16.5 Glass Plate Set for Vertical Slab Gel Systems, 14.5 cm Plate Height
Call For Price
COMPARE
16 16.5 Glass Plate Set for Vertical Slab Gel Systems, 16 cm Plate Height
Call For Price
COMPARE
19 16.5 Glass Plate Set for Vertical Slab Gel Systems, 19 cm Plate Height
Call For Price
COMPARE
22 16.5 Glass Plate Set for Vertical Slab Gel Systems, 22 cm Plate Height
Call For Price
COMPARE
28 16.5 Glass Plate Set for Vertical Slab Gel Systems, 28 cm Plate Height