Foxx Life Sciences UN/DOT Solvent Waste Systems with Rigid Tubing Ports and Hose Barbs
Provide safe HPLC waste disposal in your lab
  • Certified for UN/DOT Packaging Group II for hazardous chemicals
  • Carbon filter captures potentially harmful vapors
  • Multiple ports adapt cap to any HPLC system
  • Filter adapter elbow provides storage flexibility, especially for tight lab spaces
This solvent waste system allows easy and safe disposal of HPLC waste in the lab. The system includes a UN/DOT container with two caps; one with ports for waste collection and one to seal for transportation. Caps come with a variety of port sizes to be used with complex HPLC systems. When not in use, the ports can be plugged.
£290.70GBP / Each
£348.84GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Special Features Tubing ID
Availability Pricing
Foxx Life Sciences Waste System, UN/DOT, 70 mm Cap, 6 x 1/16" OD Tubing, 1 x 1/4" to 3/8" Hose Barb; 13.5 L WZ-06062-35 Mfr # 332-AB04-OEM
COMPARE
13.5 6 x 1/16\" OD Tubing 1 x 1/4\" to 3/8\" Hose Barb
£290.70GBP / Each
£348.84GBP (incl VAT)