Foxx Life Sciences UN/DOT Liquid Waste Disposal Containers
Safely transport hazardous liquids
  • UN/DOT rated for Group II and hazardous chemicals
  • Large top handle provides a comfortable full-hand grip
  • HDPE construction offers excellent chemical compatibility
Sturdy, rectangular UN/DOT carboys are easy to fill and ship for disposal. Safely use these single-use carboys to transfer and store liquid waste, use as waste traps with automated instruments, or as part of your HPLC system.
UN rating: 3H1/Y1.8/100
£28.80 - £32.35GBP / Each
£34.56 - £38.82GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material Overall Height (in)
Availability Pricing
Foxx Life Sciences UN/DOT Liquid Waste Container, S70 cap; 13.5 L; 1/ea WZ-06059-98 Mfr # 392-2003-OEM
COMPARE
13.5 HDPE 11
£28.80GBP / Each
£34.56GBP (incl VAT)
Foxx Life Sciences UN/DOT Liquid Waste Container, S70 cap; 20 L; 1/ea WZ-06059-99 Mfr # 392-2004-OEM
COMPARE
20 HDPE 15 1/2
£32.35GBP / Each
£38.82GBP (incl VAT)