This product is now discontinued, View Related Products
Cole-Parmer Tube Rotators
Don't compromise culture quality
  • Adjustable angle of rotation mixes samples effectively with gentle or vigorous motion
  • A variety of tube holder discs allows customization for your application
  • Integrated drip tray prevents workspace spills
These rotators mix cultures without damaging cells. Adjustable rotation angle features a scale for reproducible results, and provides gentle horizontal mixing to turbulent vertical end-over-end mixing. Operate in incubators up to 40°C and in cold rooms down to 4°C.
Choose from six tube holders to suit your application (order separately). Tools are not required to quickly and easily remove and replace tube holders. Optional dual disc adapter (00077-23) mounts two tube holders together to increase capacity.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Power (VAC)
Availability Pricing
COMPARE
20 230
Call For Price
COMPARE
20 120
Call For Price
COMPARE
40 230
Call For Price
COMPARE
40 120