Cole-Parmer® ST-200 Series Undergrad Analog Stirrers
Powerful and precision stirring in compact size
  • Built with tough aluminum casing for longevity
  • Designed to prevent accidental spillage on control panel
  • IP32 rated for water and dust resistance
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
Constructed with powerful magnets and motor, these analog stirrers provide you with stirring speeds of up to 2000 rpm and mixing capability of up to 15 L of liquid. The compact, stackable design takes up less benchtop and storage space.
For safety, all units are designed to direct spills away from the user controls. The stirrers also feature BioCote® antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.
£289.32 - £294.41GBP / Each
£347.18 - £353.29GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Material Description
Availability Pricing
COMPARE
230 Glass ceramic Stuart Undergrad Analog Stirrer, 15 x 15 cm, Ceramic; 230 VAC
£294.41GBP / Each
£353.29GBP (incl VAT)
COMPARE
120 Glass ceramic Stuart Undergrad Analog Stirrer, 6\" x 6\", Ceramic; 120 VAC
£294.41GBP / Each
£353.29GBP (incl VAT)
COMPARE
230 Stainless steel Stuart Undergrad Analog Stirrer, 15 x 15 cm, Stainless Steel; 230 VAC
£289.32GBP / Each
£347.18GBP (incl VAT)
COMPARE
120 Stainless steel Stuart Undergrad Analog Stirrer, 6\" x 6\", Stainless Steel; 120 VAC
£289.32GBP / Each
£347.18GBP (incl VAT)