Cole-Parmer® HP-300 Series Digital Round-Top Hot Plates
Simple and precise heating at your fingertips
  • Innovative fingertip control even while wearing gloves
  • Dual timer modes for more accurate control
  • Sealed casing keeps spills away from interior for maintenance-free operation
  • LCD shows set and actual temperature
  • Hot top warning lights protects you from accidental burns
  • Ceramic-coated aluminum surface offers a robust top plate with excellent heat transmission
Sleek hot plate is simple to use and provides exact heating for all your benchtop needs. Intuitive control panel has a unique pad that allows all operations to be set using just your fingertip. An LCD shows both set and real-time parameters and is lockable to prevent accidental changes during use. Optimal heating is produced by an integrated temperature sensor and dual PID control that rapidly heats plate and maintains desired temperature. Dual temperature control circuits also ensure temperature stability in case one circuit malfunctions. Safety features include user-set safety temperature limit through main control panel, hot warning light indicator, and digital error code functions for easy troubleshooting.
£477.55 - £498.41GBP / Each
£573.06 - £598.09GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Power (VAC) Top Plate Diameter (in)
Availability Pricing
COMPARE
644 110 5 19/64
£498.41GBP / Each
£598.09GBP (incl VAT)
COMPARE
644 220 5 19/64
£477.55GBP / Each
£573.06GBP (incl VAT)