Foxx Life Sciences Storage Carboys, HDPE
Secure grips make lifting, filling, and pouring easy
  • Easy-to-read metric graduations are certified to ±5% accuracy
  • Wide-mouth neck simplifies filling and cleaning
  • Ergonomic body design maximizes bench and shelf space
  • Large top handles offer full hand grip
  • Interchangeable leak-proof top connections
  • BPA-free, USP Class VI, FDA-grade resin offers excellent chemical resistance
The easy-to-grip HDPE storage carboys allow you to securely hold the carboy while lifting, filling, and pouring. The carboys feature easy-to-read metric graduation marks and molded-in material and volume identification markings. Some models offer an autoclavable PP spigot.
£54.76 - £608.53GBP / Each
£65.71 - £730.24GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Special Features
Availability Pricing
Foxx Life Sciences Carboy, HDPE, 83 mm Cap; 2.5 L WZ-06059-37 Mfr # 152-0111-OEM
COMPARE
2.5 80 mm
£54.76GBP / Each
£65.71GBP (incl VAT)
Foxx Life Sciences Carboy, HDPE, 83 mm Cap; 5 L WZ-06059-38 Mfr # 152-1111-OEM
COMPARE
5 83 mm
£90.20GBP / Each
£108.24GBP (incl VAT)
Foxx Life Sciences Carboy, HDPE, 83 mm Cap; 10 L WZ-06059-40 Mfr # 152-2111-OEM
COMPARE
10 80 mm
£111.31GBP / Each
£133.57GBP (incl VAT)
Foxx Life Sciences 152-3111-OEM-CP Carboy, HDPE, 83 mm Cap; 20 L WZ-06059-42 Mfr # 152-3111-OEM
COMPARE
20 80 mm
£123.56GBP / Each
£148.27GBP (incl VAT)
Foxx Life Sciences 152-5121-OEM-CP Carboy, HDPE, 120 mm Cap; 40 L WZ-06059-46 Mfr # 152-5121-OEM
COMPARE
40 120 mm
£191.46GBP / Each
£229.75GBP (incl VAT)
Foxx Life Sciences 152-7121-OEM-CP Carboy, HDPE, 120 mm Cap; 75 L WZ-06059-48 Mfr # 152-7121-OEM
COMPARE
75 120 mm
£608.53GBP / Each
£730.24GBP (incl VAT)