This product is now discontinued, View Related Products
Cole-Parmer Stereozoom Microscope with Large Stage 67627
LED illumination creates less heat and uses minimal energy
  • Wide magnification range from 7.5x to 50x
Larger working area is ideal for large objects and gives you room to work when dissecting. LED illumination creates little heat - ideal for temperature sensitive samples. Features top and bottom illumination with rheostat control to ensure clean image views. Inclined viewing head and interpupillary adjustment create a comfortable viewing experience. Objective lenses are high-resolution, achromatic, color-corrected, parfocalled, and parcentered.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Head Configuration
Availability Pricing
COMPARE
Binocular
Call For Price
COMPARE
Trinocular