Cole-Parmer® Essentials Square 1 qt Tall and Short Bottles, HDPE
These durable bottles are made of FDA compliant HDPE resin. Bottles have a one-quart capacity-choose either tall bottles or short bottles with handle. Both models have a tamper evident 38-mm PP cap with a PE liner that protects against leaks.
£122.91GBP / Pkg of 36
£147.49GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (oz) Material Overall Height (in)
Availability Pricing
COMPARE
32 High Density Polyethyene (HDPE) 9
£122.91GBP / Pkg of 36
£147.49GBP (incl VAT)