Cole-Parmer Single-Channel Five-Element Flame Photometer
Economical unit with tried and true capabilities inherited from higher priced models
  • Five element filter stick allows quick and easy filter selection
  • Ergonomic design with removable front tray for easy cleaning
Ideal for the testing of alkali metals in chemical manufacturing, pharmaceuticals, soil analysis, food, paper industries, and much more! Pull out or push in the filter stick to select from sodium (Na), potassium (K), lithium (Li), calcium (Ca), or barium (Ba) filters. The sample solution is aspirated into the flame, emitting a wavelength of light; the intensity of the light is proportional to the concentration of the atom in the solution. A photodetector measures the light and indicates the result on the digital display.
Features include auto ignition and flame optimization, LED indicators for selected filter, digital display with hold button, recorder output, and fail-safe flame detection with auto shutoff. Spill-containment front tray is easily removed for quick cleanup. The mixing chamber, burner head, and burner stem are disassembled without tools for easy maintenance.
Call For Price
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fuel Supply
Availability Pricing
COMPARE
Propane/butane
Call For Price
COMPARE
Natural gas