This product is now discontinued, View Related Products
Cole-Parmer Single-Channel Electronic Pipettors
Improve efficiency by saving routine dispense programs for quick access
  • Program up to six presets with function, volume, speed, and number of dispenses
  • Nine pipetting functions cover common and specialized pipetting applications
  • Adjust the large color display's brightness and background to suit any lighting conditions
This electronic pipettor is easy to use with simple plain language menus reaching only three levels in depth for quick navigation. Straightforward procedure makes calibration in the lab a snap. Pipetting functions include manual, automatic, reverse, stepped, mixing, stepped + sequence, pipette + mix, aspirate, and gel loading.
Display shows selected function, volume, piston position, selected speed, counter cycle, and battery status.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL) Min Volume (µL)
Availability Pricing
COMPARE
10 0.2
Call For Price
COMPARE
100 5
Call For Price
COMPARE
300 20
Call For Price
COMPARE
1200 50
Call For Price
COMPARE
5000 500
Call For Price
COMPARE
1000 10000