Oakton by Cole-Parmer® Self-Cleaning, Flat Surfaced pH Electrodes
No more solid buildups
  • Flat surface prevents buildups, while fluid flow cleans surface for longer life
  • Use with most meters and controllers—BNC connector and ATC element work with a variety of units
  • Double junction for difficult applications
Choose either CPVC housing or chemical-resistant Ryton® PPS housing
£160.54 - £248.74GBP / Each
£192.65 - £298.49GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
self-cleaning pH electrode, CPVC housing, 10-ft cable, BNC
£173.27GBP / Each
£207.92GBP (incl VAT)
COMPARE
self-cleaning pH electrode, CPVC housing, 100Ohm Pt RTD, mini audio
£220.29GBP / Each
£264.35GBP (incl VAT)
COMPARE
self-cleaning pH electrode, CPVC housing, 1 k Ohms RTD
£202.53GBP / Each
£243.04GBP (incl VAT)
COMPARE
self-cleaning pH electrode, CPVC housing, 3 k Ohms balco
£231.43GBP / Each
£277.72GBP (incl VAT)
COMPARE
self-cleaning pH electrode, CPVC housing, 100Ohm Pt RTD, tinned end
£221.52GBP / Each
£265.82GBP (incl VAT)
COMPARE
self-cleaning pH electrode, Ryton housing, 10-ft cable, BNC
£198.22GBP / Each
£237.86GBP (incl VAT)
COMPARE
self-cleaning pH electrode, Ryton housing, 100 Ohms RTD
£236.14GBP / Each
£283.37GBP (incl VAT)
COMPARE
self-cleaning pH electrode, PPS housing, 100 Ohms Pt RTD
£224.07GBP / Each
£268.88GBP (incl VAT)
COMPARE
self-cleaning pH electrode, Ryton housing, 1 k Ohms RTD
£248.74GBP / Each
£298.49GBP (incl VAT)
COMPARE
self-cleaning pH electrode, Ryton housing, 100 k Ohms thermistor
£160.54GBP / Each
£192.65GBP (incl VAT)
COMPARE
self-cleaning pH electrode, Ryton housing, 3 k Ohms balco
£235.27GBP / Each
£282.32GBP (incl VAT)
Showing 1- 11 of 11