Cole-Parmer® Essentials Raised Diamond Grip Nitrile Gloves
Offers superior wet and dry grip
  • Hypoallergenic
  • USDA compliant
Embossed diamond pattern provides superior wet and dry grip. Latex-free nitrile gloves resist punctures, cuts, snags, and abrasion. Silicone free—suitable for use in paint and powder coat applications. Hypoallergenic. Compliance: USDA accepted per 21CFR 177.2600 for food and drug handling, manufactured under ISO 9002 standards.
£34.57GBP / Pkg of 12
£41.48GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size Thickness (mil)
Availability Pricing
COMPARE
12 9 15
£34.57GBP / Pkg of 12
£41.48GBP (incl VAT)