Cole-Parmer® RHV-400 Series Standard Viscosity Bath
Count on constant temperatures for your stringent viscosity testing
  • Clear glass tank makes monitoring simple
  • Quickly see selected and actual values on the large, backlit display
  • Accommodates six viscometer holders
Instantly--and economically--get clear insights with this transparent viscosity bath, well-suited for testing the kinematic viscosity of petroleum, lubricants, polymer coatings, food, paint and other liquids. High-tech PID controls keep temperatures accurate and enable rapid response to changes in heat load conditions.
With a digital display and large, universal icons, the user-friendly controller features on-screen prompts and accommodates single-point calibration. You can also easily choose °F or °C data and set high/low temperature limits.
The recirculating pump optimizes temperature uniformity, while the top plate is chemical- and thermal-resistant and features lidded viscometer ports.
£3,636.50GBP / Each
£4,363.80GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
135 120 17
£3,636.50GBP / Each
£4,363.80GBP (incl VAT)